The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032522.3 | NP_115911.1 | 2 - 3 | 242 | 85 |
XM_005245546.2 | XP_005245603.1 | 2 - 3 | 300 | 85 | |
XM_011510062.2 | XP_011508364.1 | 2 - 3 | 300 | 85 | |
XM_017002558.1 | XP_016858047.1 | 2 - 3 | 300 | 85 | |
GenBank mRNA | AF318371.1 | - | 1 - 2 | 161 | 85 |
AK302696.1 | - | 2 - 3 | 217 | 85 | |
BC003116.1 | - | 1 - 2 | 203 | 85 | |
BC006315.2 | - | 1 - 2 | 203 | 85 | |
DA796002.1 | - | 2 - 3 | 242 | 85 |