Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OXNAD1
Entrez Gene ID
92106
Gene Name
oxidoreductase NAD binding domain containing 1
Gene Aliases
-
Chromosome Location
Chr.3: 16265160 - 16386979 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138381.3 NP_612390.1 6 - 7 938 114
XM_005265559.3 XP_005265616.1 6 - 7 919 114
XM_006713405.1 XP_006713468.1 6 - 7 902 114
XM_006713406.2 XP_006713469.1 7 - 8 1064 114
XM_011534230.2 XP_011532532.1 6 - 7 919 114
XM_011534232.2 XP_011532534.1 6 - 7 919 114
XM_011534233.2 XP_011532535.1 6 - 7 919 114
XM_011534234.2 XP_011532536.1 6 - 7 919 114
XM_011534235.2 XP_011532537.1 6 - 7 919 114
XM_017007485.1 XP_016862974.1 6 - 7 902 114
XM_017007486.1 XP_016862975.1 6 - 7 934 114
XM_017007487.1 XP_016862976.1 6 - 7 934 114
XM_017007488.1 XP_016862977.1 6 - 7 934 114
XM_017007489.1 XP_016862978.1 6 - 7 934 114
XM_017007490.1 XP_016862979.1 6 - 7 934 114
XM_017007491.1 XP_016862980.1 6 - 7 934 114
XM_017007492.1 XP_016862981.1 6 - 7 673 114
XM_017007493.1 XP_016862982.1 7 - 8 1064 114
XM_017007494.1 XP_016862983.1 7 - 8 1064 114
XM_017007495.1 XP_016862984.1 7 - 8 1061 114
XM_017007496.1 XP_016862985.1 7 - 8 1061 114
GenBank mRNA AB209556.1 - 4 - 5 432 114
AK298829.1 - 5 - 6 493 114
AK307258.1 - 5 - 6 801 114
AL583453.3 - 6 - 7 897 114
AL832787.1 - 2 - 3 240 114
BC008322.1 - 6 - 7 865 114
DQ364598.1 - 6 - 7 896 114

View Full Product Details