The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278298.1 | NP_001265227.1 | 12 - 13 | 4719 | 63 |
NM_153264.6 | NP_694996.5 | 12 - 13 | 4719 | 63 | |
NR_022012.2 | 12 - 13 | 4719 | 63 | ||
XM_011512621.2 | XP_011510923.1 | 12 - 13 | 4593 | 63 | |
XM_011512622.2 | XP_011510924.1 | 12 - 13 | 4593 | 63 | |
XM_011512623.2 | XP_011510925.1 | 12 - 13 | 4593 | 63 | |
GenBank mRNA | AM906080.1 | - | 6 - 7 | 1546 | 63 |
AM906081.1 | - | 4 - 5 | 661 | 63 | |
EU085556.1 | - | 12 - 13 | 4719 | 63 |