Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL6A5
Entrez Gene ID
256076
Gene Name
collagen type VI alpha 5
Gene Aliases
COL29A1, VWA4
Chromosome Location
Chr.3: 130345516 - 130484846 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278298.1 NP_001265227.1 13 - 14 4777 87
NM_153264.6 NP_694996.5 13 - 14 4777 87
NR_022012.2 13 - 14 4777 87
XM_011512621.2 XP_011510923.1 13 - 14 4651 87
XM_011512622.2 XP_011510924.1 13 - 14 4651 87
XM_011512623.2 XP_011510925.1 13 - 14 4651 87
GenBank mRNA AM906081.1 - 5 - 6 719 87
EU085556.1 - 13 - 14 4777 87

View Full Product Details