The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278298.1 | NP_001265227.1 | 7 - 8 | 3483 | 150 |
NM_153264.6 | NP_694996.5 | 7 - 8 | 3483 | 150 | |
NR_022012.2 | 7 - 8 | 3483 | 150 | ||
XM_011512621.2 | XP_011510923.1 | 7 - 8 | 3357 | 150 | |
XM_011512622.2 | XP_011510924.1 | 7 - 8 | 3357 | 150 | |
XM_011512623.2 | XP_011510925.1 | 7 - 8 | 3357 | 150 | |
GenBank mRNA | AM906080.1 | - | 1 - 2 | 310 | 150 |
EU085556.1 | - | 7 - 8 | 3483 | 150 |