The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015941.3 | NP_057025.2 | 8 - 9 | 989 | 72 |
NM_213619.2 | NP_998784.1 | 7 - 8 | 935 | 72 | |
NM_213620.2 | NP_998785.1 | 8 - 9 | 883 | 72 | |
XM_006716455.2 | XP_006716518.1 | 7 - 8 | 820 | 72 | |
XM_011517542.1 | XP_011515844.1 | 5 - 6 | 460 | 72 | |
GenBank mRNA | AF112204.1 | - | 8 - 9 | 919 | 72 |
AF113222.1 | - | 7 - 8 | 901 | 72 | |
AF125105.1 | - | - | 846 | 72 | |
AF298777.1 | - | 7 - 8 | 676 | 72 | |
AK022345.1 | - | 8 - 9 | 940 | 72 | |
AK094839.1 | - | 7 - 8 | 900 | 72 | |
AK098305.1 | - | 8 - 9 | 828 | 72 | |
AK303229.1 | - | 8 - 9 | 893 | 72 | |
AK308204.1 | - | 8 - 9 | 799 | 72 | |
BC025275.1 | - | 8 - 9 | 912 | 72 |