The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014648.3 | NP_055463.1 | 14 - 15 | 2178 | 89 |
XM_005247914.3 | XP_005247971.1 | 14 - 15 | 1977 | 89 | |
XM_005247915.3 | XP_005247972.1 | 14 - 15 | 1947 | 89 | |
XM_005247916.3 | XP_005247973.1 | 15 - 16 | 2119 | 89 | |
XM_005247917.3 | XP_005247974.1 | 14 - 15 | 1908 | 89 | |
XM_005247918.3 | XP_005247975.1 | 14 - 15 | 2395 | 89 | |
XM_017007540.1 | XP_016863029.1 | 14 - 15 | 2024 | 89 | |
GenBank mRNA | AB014575.1 | - | 14 - 15 | 1885 | 89 |
AK023138.1 | - | 14 - 15 | 1946 | 89 | |
AK023389.1 | - | 14 - 15 | 1987 | 89 | |
AK294967.1 | - | 3 - 4 | 241 | 89 | |
AK303942.1 | - | 1 - 2 | 381 | 89 | |
AY227651.1 | - | 14 - 15 | 1924 | 89 | |
AY227652.1 | - | 1 - 2 | 89 | 89 | |
AY227653.1 | - | 5 - 6 | 941 | 89 | |
AY227654.1 | - | 14 - 15 | 1949 | 89 | |
BC039018.1 | - | 14 - 15 | 1939 | 89 | |
BC063882.1 | - | 14 - 15 | 1931 | 89 |