The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014648.3 | NP_055463.1 | 15 - 16 | 2210 | 141 |
XM_005247914.3 | XP_005247971.1 | 15 - 16 | 2009 | 141 | |
XM_005247915.3 | XP_005247972.1 | 15 - 16 | 1979 | 141 | |
XM_005247916.3 | XP_005247973.1 | 16 - 17 | 2151 | 141 | |
XM_005247917.3 | XP_005247974.1 | 15 - 16 | 1940 | 141 | |
XM_005247918.3 | XP_005247975.1 | 15 - 16 | 2427 | 141 | |
XM_017007540.1 | XP_016863029.1 | 15 - 16 | 2056 | 141 | |
GenBank mRNA | AB014575.1 | - | 15 - 16 | 1917 | 141 |
AK023138.1 | - | 15 - 16 | 1978 | 141 | |
AK023389.1 | - | 15 - 16 | 2019 | 141 | |
AK303942.1 | - | 2 - 3 | 413 | 141 | |
AY227651.1 | - | 15 - 16 | 1956 | 141 | |
AY227652.1 | - | 2 - 3 | 121 | 141 | |
AY227653.1 | - | 6 - 7 | 973 | 141 | |
AY227654.1 | - | 15 - 16 | 1981 | 141 | |
BC039018.1 | - | 15 - 16 | 1971 | 141 | |
BC063882.1 | - | 15 - 16 | 1963 | 141 |