Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIF14
Entrez Gene ID
9928
Gene Name
kinesin family member 14
Gene Aliases
MKS12
Chromosome Location
Chr.1: 200551497 - 200620791 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001305792.1 NP_001292721.1 14 - 15 2841 106
NM_014875.2 NP_055690.1 16 - 17 3253 106
XM_011510231.2 XP_011508533.1 17 - 18 3347 106
XM_011510232.2 XP_011508534.1 16 - 17 3009 106
XM_011510233.2 XP_011508535.1 17 - 18 3140 106
XM_011510235.2 XP_011508537.1 17 - 18 2852 106
XM_011510236.1 XP_011508538.1 15 - 16 3136 106
XM_017003005.1 XP_016858494.1 15 - 16 3430 106
XM_017003006.1 XP_016858495.1 16 - 17 3038 106
XM_017003007.1 XP_016858496.1 16 - 17 2565 106
GenBank mRNA BC098582.1 - 6 - 7 846 106
BC113742.1 - 15 - 16 2892 106
D26361.1 - 16 - 17 3253 106

View Full Product Details