Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIF14
Entrez Gene ID
9928
Gene Name
kinesin family member 14
Gene Aliases
MKS12
Chromosome Location
Chr.1: 200551497 - 200620791 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001305792.1 NP_001292721.1 17 - 18 3275 154
NM_014875.2 NP_055690.1 19 - 20 3687 154
XM_011510231.2 XP_011508533.1 20 - 21 3781 154
XM_011510232.2 XP_011508534.1 19 - 20 3443 154
XM_011510233.2 XP_011508535.1 20 - 21 3574 154
XM_011510235.2 XP_011508537.1 20 - 21 3286 154
XM_011510236.1 XP_011508538.1 18 - 19 3570 154
XM_017003005.1 XP_016858494.1 18 - 19 3864 154
XM_017003006.1 XP_016858495.1 19 - 20 3472 154
XM_017003007.1 XP_016858496.1 19 - 20 2999 154
GenBank mRNA BC098582.1 - 9 - 10 1280 154
BC113742.1 - 18 - 19 3326 154
D26361.1 - 19 - 20 3687 154

View Full Product Details