Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIF14
Entrez Gene ID
9928
Gene Name
kinesin family member 14
Gene Aliases
MKS12
Chromosome Location
Chr.1: 200551497 - 200620791 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001305792.1 NP_001292721.1 18 - 19 3367 108
NM_014875.2 NP_055690.1 20 - 21 3779 108
XM_011510231.2 XP_011508533.1 21 - 22 3873 108
XM_011510232.2 XP_011508534.1 20 - 21 3535 108
XM_011510233.2 XP_011508535.1 21 - 22 3666 108
XM_011510235.2 XP_011508537.1 21 - 22 3378 108
XM_011510236.1 XP_011508538.1 19 - 20 3662 108
XM_017003005.1 XP_016858494.1 19 - 20 3956 108
XM_017003006.1 XP_016858495.1 20 - 21 3564 108
XM_017003007.1 XP_016858496.1 20 - 21 3091 108
GenBank mRNA BC098582.1 - 10 - 11 1372 108
BC113742.1 - 19 - 20 3418 108
D26361.1 - 20 - 21 3779 108

View Full Product Details