The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001305792.1 | NP_001292721.1 | 26 - 27 | 4457 | 123 |
NM_014875.2 | NP_055690.1 | 28 - 29 | 4869 | 123 | |
XM_011510231.2 | XP_011508533.1 | 29 - 30 | 4963 | 123 | |
XM_011510232.2 | XP_011508534.1 | 28 - 29 | 4625 | 123 | |
XM_011510233.2 | XP_011508535.1 | 29 - 30 | 4756 | 123 | |
XM_011510235.2 | XP_011508537.1 | 29 - 30 | 4468 | 123 | |
XM_011510236.1 | XP_011508538.1 | 27 - 28 | 4752 | 123 | |
XM_017003005.1 | XP_016858494.1 | 27 - 28 | 5046 | 123 | |
XM_017003006.1 | XP_016858495.1 | 28 - 29 | 4654 | 123 | |
XM_017003007.1 | XP_016858496.1 | 28 - 29 | 4181 | 123 | |
GenBank mRNA | BC098582.1 | - | 18 - 19 | 2462 | 123 |
BC113742.1 | - | 27 - 28 | 4508 | 123 | |
D26361.1 | - | 28 - 29 | 4869 | 123 |