Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KIF14
Entrez Gene ID
9928
Gene Name
kinesin family member 14
Gene Aliases
MKS12
Chromosome Location
Chr.1: 200551497 - 200620791 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001305792.1 NP_001292721.1 7 - 8 1887 106
NM_014875.2 NP_055690.1 9 - 10 2299 106
XM_011510231.2 XP_011508533.1 10 - 11 2393 106
XM_011510232.2 XP_011508534.1 9 - 10 2055 106
XM_011510233.2 XP_011508535.1 10 - 11 2186 106
XM_011510235.2 XP_011508537.1 10 - 11 1898 106
XM_011510236.1 XP_011508538.1 8 - 9 2182 106
XM_017003005.1 XP_016858494.1 8 - 9 2476 106
XM_017003006.1 XP_016858495.1 9 - 10 2084 106
XM_017003007.1 XP_016858496.1 9 - 10 1611 106
GenBank mRNA BC113742.1 - 8 - 9 1938 106
D26361.1 - 9 - 10 2299 106

View Full Product Details