Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMG7
Entrez Gene ID
9887
Gene Name
SMG7, nonsense mediated mRNA decay factor
Gene Aliases
C1orf16, EST1C, SGA56M
Chromosome Location
Chr.1: 183471962 - 183554193 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201568.2 NP_963862.1 14 - 15 1955 159
NM_201569.2 NP_963863.2 14 - 15 1955 159
XM_006711676.3 XP_006711739.1 15 - 16 2503 159
XM_006711678.3 XP_006711741.1 14 - 15 1824 159
XM_011510206.2 XP_011508508.1 17 - 18 2818 159
XM_011510209.2 XP_011508511.1 17 - 18 2818 159
XM_011510210.2 XP_011508512.1 13 - 14 1794 159
XM_017002966.1 XP_016858455.1 16 - 17 2095 159
XM_017002967.1 XP_016858456.1 16 - 17 2103 159
XM_017002968.1 XP_016858457.1 13 - 14 1798 159
XM_017002971.1 XP_016858460.1 14 - 15 2017 159
GenBank mRNA AK056035.1 - 14 - 15 1824 159
AK074297.1 - 14 - 15 1812 159
AK091477.1 - 13 - 14 1792 159
AK297207.1 - 15 - 16 1987 159
AK307394.1 - 14 - 15 1816 159
AL833675.1 - 2 - 3 338 159
BC036381.1 - 14 - 15 1823 159
BC052565.1 - 7 - 8 950 159
D87437.2 - 14 - 15 1799 159

View Full Product Details