The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001292008.1 | NP_001278937.1 | 6 - 7 | 695 | 152 |
NM_003144.4 | NP_003135.2 | 6 - 7 | 899 | 152 | |
GenBank mRNA | AF156965.1 | - | 6 - 7 | 816 | 152 |
AK075562.1 | - | 5 - 6 | 683 | 152 | |
AK222762.1 | - | 6 - 7 | 767 | 152 | |
AK222809.1 | - | 6 - 7 | 767 | 152 | |
AK291550.1 | - | 6 - 7 | 767 | 152 | |
AL550910.3 | - | 6 - 7 | 556 | 152 | |
BC007710.2 | - | 6 - 7 | 792 | 152 | |
BT007387.1 | - | 6 - 7 | 705 | 152 | |
CR456961.1 | - | 6 - 7 | 705 | 152 | |
CR936719.1 | - | 5 - 6 | 758 | 152 | |
Z12830.1 | - | 6 - 7 | 734 | 152 |