Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NOL11
Entrez Gene ID
25926
Gene Name
nucleolar protein 11
Gene Aliases
-
Chromosome Location
Chr.17: 67717833 - 67744208 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303272.1 NP_001290201.1 15 - 16 1986 129
NM_015462.4 NP_056277.2 16 - 17 2044 129
GenBank mRNA AK023702.1 - 16 - 17 1934 129
AK025390.1 - 4 - 5 1548 129
AK299483.1 - 15 - 16 1876 129
AL110271.1 - 13 - 14 1634 129
AY598333.1 - 16 - 17 1929 129
BC001726.2 - 8 - 9 998 129
BC064404.1 - 16 - 17 1935 129

View Full Product Details