The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303272.1 | NP_001290201.1 | 8 - 9 | 1110 | 81 |
NM_015462.4 | NP_056277.2 | 9 - 10 | 1168 | 81 | |
GenBank mRNA | AK023702.1 | - | 9 - 10 | 1058 | 81 |
AK299483.1 | - | 8 - 9 | 1000 | 81 | |
AL110271.1 | - | 6 - 7 | 763 | 81 | |
AY598333.1 | - | 9 - 10 | 1053 | 81 | |
BC001726.2 | - | 1 - 2 | 122 | 81 | |
BC064404.1 | - | 9 - 10 | 1059 | 81 |