The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164391.1 | NP_001157863.1 | 14 - 15 | 1528 | 108 |
NM_001164392.1 | NP_001157864.1 | 12 - 13 | 1437 | 108 | |
NM_001164393.1 | NP_001157865.1 | 11 - 12 | 1327 | 108 | |
NM_007358.3 | NP_031384.1 | 13 - 14 | 1608 | 108 | |
XM_011541016.1 | XP_011539318.1 | 13 - 14 | 1564 | 108 | |
XM_011541019.1 | XP_011539321.1 | 13 - 14 | 1356 | 108 | |
XM_017000676.1 | XP_016856165.1 | 13 - 14 | 1463 | 108 | |
XM_017000677.1 | XP_016856166.1 | 12 - 13 | 1393 | 108 | |
XM_017000678.1 | XP_016856167.1 | 13 - 14 | 1346 | 108 | |
XM_017000679.1 | XP_016856168.1 | 12 - 13 | 1440 | 108 | |
GenBank mRNA | AF072814.1 | - | 12 - 13 | 1306 | 108 |
AF073293.1 | - | 3 - 4 | 810 | 108 | |
AF086908.1 | - | 12 - 13 | 1409 | 108 | |
AJ010014.1 | - | 13 - 14 | 1559 | 108 | |
AK290318.1 | - | 13 - 14 | 1560 | 108 | |
AK302776.1 | - | 14 - 15 | 1466 | 108 | |
AK302900.1 | - | 11 - 12 | 1279 | 108 | |
BC010013.2 | - | 12 - 13 | 1361 | 108 |