The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164391.1 | NP_001157863.1 | 5 - 6 | 588 | 124 |
NM_001164392.1 | NP_001157864.1 | 4 - 5 | 668 | 124 | |
NM_001164393.1 | NP_001157865.1 | 2 - 3 | 387 | 124 | |
NM_007358.3 | NP_031384.1 | 4 - 5 | 668 | 124 | |
XM_011541016.1 | XP_011539318.1 | 4 - 5 | 624 | 124 | |
XM_011541017.2 | XP_011539319.1 | 4 - 5 | 684 | 124 | |
XM_011541019.1 | XP_011539321.1 | 5 - 6 | 540 | 124 | |
XM_017000676.1 | XP_016856165.1 | 4 - 5 | 523 | 124 | |
XM_017000677.1 | XP_016856166.1 | 4 - 5 | 624 | 124 | |
XM_017000678.1 | XP_016856167.1 | 5 - 6 | 577 | 124 | |
XM_017000679.1 | XP_016856168.1 | 4 - 5 | 624 | 124 | |
GenBank mRNA | AF072814.1 | - | 3 - 4 | 366 | 124 |
AJ010014.1 | - | 4 - 5 | 619 | 124 | |
AK290318.1 | - | 4 - 5 | 620 | 124 | |
AK302776.1 | - | 5 - 6 | 527 | 124 | |
AK302900.1 | - | 2 - 3 | 339 | 124 | |
BC001897.1 | - | 4 - 5 | 587 | 124 | |
BC010013.2 | - | 4 - 5 | 592 | 124 |