Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTF2
Entrez Gene ID
22823
Gene Name
metal response element binding transcription factor 2
Gene Aliases
M96, PCL2, TDRD19A, dJ976O13.2
Chromosome Location
Chr.1: 93079219 - 93139081 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164391.1 NP_001157863.1 7 - 8 845 78
NM_001164392.1 NP_001157864.1 6 - 7 925 78
NM_001164393.1 NP_001157865.1 4 - 5 644 78
NM_007358.3 NP_031384.1 6 - 7 925 78
XM_011541016.1 XP_011539318.1 6 - 7 881 78
XM_011541017.2 XP_011539319.1 6 - 7 941 78
XM_011541019.1 XP_011539321.1 7 - 8 797 78
XM_017000676.1 XP_016856165.1 6 - 7 780 78
XM_017000677.1 XP_016856166.1 6 - 7 881 78
XM_017000678.1 XP_016856167.1 7 - 8 834 78
XM_017000679.1 XP_016856168.1 6 - 7 881 78
GenBank mRNA AF072814.1 - 5 - 6 623 78
AF086908.1 - 6 - 7 850 78
AJ010014.1 - 6 - 7 876 78
AK290318.1 - 6 - 7 877 78
AK302776.1 - 7 - 8 784 78
AK302900.1 - 4 - 5 596 78
BC001897.1 - 6 - 7 844 78
BC010013.2 - 6 - 7 849 78

View Full Product Details