The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039707.1 | NP_001034796.1 | 3 - 4 | 494 | 113 |
XM_005266050.3 | XP_005266107.1 | 3 - 4 | 448 | 113 | |
GenBank mRNA | AL559847.3 | - | 3 - 4 | 428 | 113 |
AY349357.1 | - | 2 - 3 | 115 | 113 |