Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SDCCAG3
Entrez Gene ID
10807
Gene Name
serologically defined colon cancer antigen 3
Gene Aliases
NY-CO-3
Chromosome Location
Chr.9: 136401922 - 136410602 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039707.1 NP_001034796.1 5 - 6 1025 113
NM_001039708.1 NP_001034797.1 3 - 4 806 113
NM_006643.3 NP_006634.3 4 - 5 956 113
XM_005266050.3 XP_005266107.1 5 - 6 979 113
XM_005266051.3 XP_005266108.1 4 - 5 870 113
XM_011518157.2 XP_011516459.1 4 - 5 809 113
XM_017014218.1 XP_016869707.1 4 - 5 1126 113
XM_017014219.1 XP_016869708.1 4 - 5 1126 113
GenBank mRNA AF039688.1 - 1 - 2 93 113
AK001296.1 - 3 - 4 585 113
AY349357.1 - 4 - 5 646 113
BC014515.1 - 4 - 5 823 113

View Full Product Details