Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COPB2
Entrez Gene ID
9276
Gene Name
coatomer protein complex subunit beta 2
Gene Aliases
beta'-COP
Chromosome Location
Chr.3: 139355595 - 139389680 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004766.2 NP_004757.1 14 - 15 1805 88
NR_023350.1 14 - 15 1884 88
XM_017007512.1 XP_016863001.1 15 - 16 1990 88
XM_017007513.1 XP_016863002.1 13 - 14 1551 88
GenBank mRNA AK302943.1 - 14 - 15 1718 88
BC000326.2 - 14 - 15 1751 88
X70476.1 - 14 - 15 1743 88

View Full Product Details