Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PET100
Entrez Gene ID
100131801
Gene Name
PET100 homolog
Gene Aliases
C19orf79
Chromosome Location
Chr.19: 7629785 - 7631624 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_033242.1 3 - 4 192 99
GenBank mRNA CB119056.1 - 3 - 4 183 99

View Full Product Details