The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286559.1 | NP_001273488.1 | 5 - 6 | 1945 | 99 |
NM_144705.3 | NP_653306.1 | 5 - 6 | 1242 | 99 | |
XM_011510670.1 | XP_011508972.1 | 5 - 6 | 5031 | 99 | |
GenBank mRNA | AK097438.1 | - | 5 - 6 | 1945 | 99 |
BC021716.2 | - | 5 - 6 | 1188 | 99 |