The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005233.5 | NP_005224.2 | 14 - 15 | 2718 | 69 |
XM_005264715.2 | XP_005264772.1 | 14 - 15 | 2640 | 69 | |
XM_005264716.2 | XP_005264773.1 | 14 - 15 | 2713 | 69 | |
GenBank mRNA | AF213459.1 | - | 14 - 15 | 2718 | 69 |
AK291411.1 | - | 14 - 15 | 2551 | 69 | |
BC063282.1 | - | 14 - 15 | 2622 | 69 | |
M83941.1 | - | 14 - 15 | 2593 | 69 |