The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003105.5 | NP_003096.1 | 40 - 41 | 5551 | 86 |
XM_011542963.2 | XP_011541265.1 | 39 - 40 | 5312 | 86 | |
XM_011542965.2 | XP_011541267.1 | 34 - 35 | 4460 | 86 | |
XM_011542967.2 | XP_011541269.1 | 20 - 21 | 2492 | 86 | |
XM_017018169.1 | XP_016873658.1 | 40 - 41 | 5258 | 86 | |
XM_017018170.1 | XP_016873659.1 | 38 - 39 | 4934 | 86 | |
XM_017018172.1 | XP_016873661.1 | 26 - 27 | 3423 | 86 | |
GenBank mRNA | AK294404.1 | - | 11 - 12 | 1689 | 86 |
AK295090.1 | - | 17 - 18 | 2341 | 86 | |
AK295878.1 | - | 20 - 21 | 2492 | 86 | |
AK302589.1 | - | 17 - 18 | 2163 | 86 | |
AK316340.1 | - | 11 - 12 | 1271 | 86 | |
BC137171.1 | - | 40 - 41 | 5474 | 86 | |
Y08110.1 | - | 40 - 41 | 5502 | 86 |