The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003105.5 | NP_003096.1 | 45 - 46 | 6297 | 113 |
XM_011542963.2 | XP_011541265.1 | 44 - 45 | 6058 | 113 | |
XM_011542965.2 | XP_011541267.1 | 39 - 40 | 5206 | 113 | |
XM_011542967.2 | XP_011541269.1 | 25 - 26 | 3238 | 113 | |
XM_017018169.1 | XP_016873658.1 | 45 - 46 | 6004 | 113 | |
XM_017018170.1 | XP_016873659.1 | 43 - 44 | 5680 | 113 | |
XM_017018172.1 | XP_016873661.1 | 31 - 32 | 4169 | 113 | |
GenBank mRNA | AK294404.1 | - | 16 - 17 | 2435 | 113 |
AK295090.1 | - | 22 - 23 | 3087 | 113 | |
AK295878.1 | - | 25 - 26 | 3238 | 113 | |
AK302589.1 | - | 22 - 23 | 2909 | 113 | |
AK316340.1 | - | 16 - 17 | 2017 | 113 | |
BC137171.1 | - | 45 - 46 | 6220 | 113 | |
Y08110.1 | - | 45 - 46 | 6248 | 113 |