Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NTRK3
Entrez Gene ID
4916
Gene Name
neurotrophic receptor tyrosine kinase 3
Gene Aliases
GP145-TrkC, TRKC, gp145(trkC)
Chromosome Location
Chr.15: 87859749 - 88256796 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007156.2 NP_001007157.1 11 - 12 1539 103
NM_001012338.2 NP_001012338.1 11 - 12 1539 103
NM_001320134.1 NP_001307063.1 9 - 10 1249 103
NM_001320135.1 NP_001307064.1 9 - 10 1140 103
NM_002530.3 NP_002521.2 11 - 12 1539 103
XM_006720543.3 XP_006720606.1 9 - 10 1848 103
XM_006720545.3 XP_006720608.1 9 - 10 1848 103
XM_006720548.3 XP_006720611.1 9 - 10 1848 103
XM_006720549.3 XP_006720612.1 9 - 10 1848 103
XM_011521637.2 XP_011519939.1 9 - 10 1848 103
XM_011521638.2 XP_011519940.1 9 - 10 1848 103
XM_017022240.1 XP_016877729.1 10 - 11 1719 103
XM_017022241.1 XP_016877730.1 9 - 10 1204 103
XM_017022242.1 XP_016877731.1 9 - 10 1848 103
XM_017022243.1 XP_016877732.1 9 - 10 1200 103
XM_017022244.1 XP_016877733.1 9 - 10 1102 103
XM_017022245.1 XP_016877734.1 10 - 11 1157 103
XM_017022246.1 XP_016877735.1 9 - 10 1258 103
XM_017022249.1 XP_016877738.1 9 - 10 1848 103
XM_017022250.1 XP_016877739.1 9 - 10 1848 103
XM_017022253.1 XP_016877742.1 9 - 10 1848 103
XM_017022254.1 XP_016877743.1 9 - 10 1848 103
GenBank mRNA AK302504.1 - 9 - 10 1099 103
BC013693.1 - 9 - 10 1238 103
BT007291.1 - 9 - 10 1233 103
KJ534904.1 - 9 - 10 1233 103
KJ534905.1 - 9 - 10 1233 103
S76475.1 - 9 - 10 1248 103
S76476.1 - 9 - 10 1248 103
U05012.1 - 10 - 11 1388 103

View Full Product Details