Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NTRK3
Entrez Gene ID
4916
Gene Name
neurotrophic receptor tyrosine kinase 3
Gene Aliases
GP145-TrkC, TRKC, gp145(trkC)
Chromosome Location
Chr.15: 87859749 - 88256796 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007156.2 NP_001007157.1 12 - 13 1595 65
NM_001012338.2 NP_001012338.1 12 - 13 1595 65
NM_001243101.1 NP_001230030.1 11 - 12 1571 65
NM_001320134.1 NP_001307063.1 10 - 11 1305 65
NM_001320135.1 NP_001307064.1 10 - 11 1196 65
NM_002530.3 NP_002521.2 12 - 13 1595 65
XM_006720543.3 XP_006720606.1 10 - 11 1904 65
XM_006720544.3 XP_006720607.1 9 - 10 1880 65
XM_006720545.3 XP_006720608.1 10 - 11 1904 65
XM_006720548.3 XP_006720611.1 10 - 11 1904 65
XM_006720549.3 XP_006720612.1 10 - 11 1904 65
XM_006720550.3 XP_006720613.1 9 - 10 1880 65
XM_011521637.2 XP_011519939.1 10 - 11 1904 65
XM_011521638.2 XP_011519940.1 10 - 11 1904 65
XM_017022240.1 XP_016877729.1 11 - 12 1775 65
XM_017022241.1 XP_016877730.1 10 - 11 1260 65
XM_017022242.1 XP_016877731.1 10 - 11 1904 65
XM_017022243.1 XP_016877732.1 10 - 11 1256 65
XM_017022244.1 XP_016877733.1 10 - 11 1158 65
XM_017022245.1 XP_016877734.1 11 - 12 1213 65
XM_017022246.1 XP_016877735.1 10 - 11 1314 65
XM_017022247.1 XP_016877736.1 9 - 10 1880 65
XM_017022248.1 XP_016877737.1 9 - 10 1880 65
XM_017022249.1 XP_016877738.1 10 - 11 1904 65
XM_017022250.1 XP_016877739.1 10 - 11 1904 65
XM_017022251.1 XP_016877740.1 3 - 4 275 65
XM_017022252.1 XP_016877741.1 2 - 3 251 65
XM_017022253.1 XP_016877742.1 10 - 11 1904 65
XM_017022254.1 XP_016877743.1 10 - 11 1904 65
GenBank mRNA AF052184.1 - 1 - 2 57 65
AK297160.1 - 9 - 10 1289 65
AK302504.1 - 10 - 11 1155 65
AK313646.1 - 9 - 10 1290 65
BC013693.1 - 10 - 11 1294 65
BT007291.1 - 10 - 11 1289 65
KJ534904.1 - 10 - 11 1289 65
KJ534905.1 - 10 - 11 1289 65
KJ534906.1 - 9 - 10 1265 65
KJ535052.1 - 9 - 10 1265 65
S76475.1 - 10 - 11 1304 65
S76476.1 - 10 - 11 1304 65
U05012.1 - 11 - 12 1444 65

View Full Product Details