Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMYD3
Entrez Gene ID
64754
Gene Name
SET and MYND domain containing 3
Gene Aliases
KMT3E, ZMYND1, ZNFN3A1, bA74P14.1
Chromosome Location
Chr.1: 245749340 - 246507342 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167740.1 NP_001161212.1 1 - 2 294 102
XM_011544253.1 XP_011542555.1 1 - 2 270 102
GenBank mRNA AB057595.1 - 1 - 2 264 102
AK289605.1 - 1 - 2 270 102
BC031010.1 - 1 - 2 209 102
BC107725.1 - 1 - 2 268 102
BI916760.1 - 1 - 2 186 102
BP231249.1 - 1 - 2 293 102

View Full Product Details