Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PHLDB1
Entrez Gene ID
23187
Gene Name
pleckstrin homology like domain family B member 1
Gene Aliases
LL5A
Chromosome Location
Chr.11: 118606296 - 118658038 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144758.2 NP_001138230.1 21 - 22 4088 83
NM_015157.3 NP_055972.1 22 - 23 4311 83
XM_005271468.3 XP_005271525.1 22 - 23 4902 83
XM_005271470.3 XP_005271527.1 21 - 22 4857 83
XM_005271472.3 XP_005271529.1 21 - 22 4761 83
XM_005271473.3 XP_005271530.1 20 - 21 4716 83
XM_005277680.3 XP_005277737.1 25 - 26 5343 83
XM_006718796.3 XP_006718859.1 25 - 26 4532 83
XM_006718797.3 XP_006718860.1 24 - 25 5202 83
XM_006718798.3 XP_006718861.1 23 - 24 5031 83
XM_006718799.3 XP_006718862.1 22 - 23 4986 83
XM_006718801.3 XP_006718864.1 22 - 23 4890 83
XM_006718802.3 XP_006718865.1 21 - 22 4845 83
XM_011542703.2 XP_011541005.1 25 - 26 5346 83
XM_011542705.2 XP_011541007.1 24 - 25 5301 83
XM_011542707.2 XP_011541009.1 24 - 25 5217 83
XM_011542708.2 XP_011541010.1 25 - 26 4535 83
XM_011542709.2 XP_011541011.1 25 - 26 4845 83
XM_011542710.2 XP_011541012.1 24 - 25 5175 83
XM_011542711.2 XP_011541013.1 22 - 23 4252 83
XM_011542713.2 XP_011541015.1 22 - 23 4246 83
XM_017017407.1 XP_016872896.1 24 - 25 5214 83
XM_017017410.1 XP_016872899.1 23 - 24 5073 83
GenBank mRNA AB014538.2 - 21 - 22 4065 83
AB094090.1 - 22 - 23 4311 83

View Full Product Details