Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHLDB1
Entrez Gene ID
23187
Gene Name
pleckstrin homology like domain family B member 1
Gene Aliases
LL5A
Chromosome Location
Chr.11: 118606296 - 118658038 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144758.2 NP_001138230.1 3 - 4 315 88
NM_001144759.2 NP_001138231.1 3 - 4 315 88
NM_015157.3 NP_055972.1 4 - 5 538 88
XM_005271468.3 XP_005271525.1 3 - 4 1084 88
XM_005271469.3 XP_005271526.1 3 - 4 1084 88
XM_005271470.3 XP_005271527.1 3 - 4 1084 88
XM_005271471.3 XP_005271528.1 3 - 4 1084 88
XM_005271472.3 XP_005271529.1 3 - 4 1084 88
XM_005271473.3 XP_005271530.1 3 - 4 1084 88
XM_005271474.4 XP_005271531.1 3 - 4 1084 88
XM_005271475.3 XP_005271532.1 3 - 4 1084 88
XM_005277680.3 XP_005277737.1 3 - 4 1084 88
XM_006718796.3 XP_006718859.1 3 - 4 273 88
XM_006718797.3 XP_006718860.1 3 - 4 1084 88
XM_006718798.3 XP_006718861.1 3 - 4 1084 88
XM_006718799.3 XP_006718862.1 3 - 4 1084 88
XM_006718800.3 XP_006718863.1 3 - 4 1084 88
XM_006718801.3 XP_006718864.1 3 - 4 1084 88
XM_006718802.3 XP_006718865.1 3 - 4 1084 88
XM_006718803.3 XP_006718866.1 3 - 4 1084 88
XM_011542703.2 XP_011541005.1 3 - 4 1084 88
XM_011542704.2 XP_011541006.1 3 - 4 1084 88
XM_011542705.2 XP_011541007.1 3 - 4 1084 88
XM_011542706.2 XP_011541008.1 3 - 4 1084 88
XM_011542707.2 XP_011541009.1 3 - 4 1084 88
XM_011542708.2 XP_011541010.1 3 - 4 273 88
XM_011542709.2 XP_011541011.1 3 - 4 583 88
XM_011542710.2 XP_011541012.1 3 - 4 1084 88
XM_011542714.2 XP_011541016.1 3 - 4 1084 88
XM_017017407.1 XP_016872896.1 3 - 4 1084 88
XM_017017408.1 XP_016872897.1 3 - 4 1084 88
XM_017017409.1 XP_016872898.1 3 - 4 1084 88
XM_017017410.1 XP_016872899.1 3 - 4 1084 88
XM_017017411.1 XP_016872900.1 3 - 4 1084 88
GenBank mRNA AB014538.2 - 3 - 4 292 88
AB094090.1 - 4 - 5 538 88
AK074070.2 - 3 - 4 264 88
AK125783.1 - 3 - 4 324 88
AK295819.1 - 3 - 4 273 88
AL832373.1 - 3 - 4 272 88
BC098586.1 - 3 - 4 292 88
BX489377.1 - 3 - 4 316 88
CR749783.1 - 4 - 5 451 88

View Full Product Details