Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SYNCRIP
Entrez Gene ID
10492
Gene Name
synaptotagmin binding cytoplasmic RNA interacting protein
Gene Aliases
GRY-RBP, GRYRBP, HNRNPQ, HNRPQ1, NSAP1, PP68, hnRNP-Q
Chromosome Location
Chr.6: 85607784 - 85643862 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159674.1 NP_001153146.1 2 - 3 253 126
NM_001159675.1 NP_001153147.1 1 - 2 159 126
NM_001159676.1 NP_001153148.1 1 - 2 159 126
NM_001159677.1 NP_001153149.1 2 - 3 688 126
NM_006372.4 NP_006363.4 2 - 3 688 126
XM_005248635.4 XP_005248692.1 2 - 3 309 126
XM_005248637.2 XP_005248694.1 2 - 3 272 126
XM_017010176.1 XP_016865665.1 2 - 3 309 126
XM_017010177.1 XP_016865666.1 2 - 3 309 126
XM_017010178.1 XP_016865667.1 2 - 3 269 126
XM_017010179.1 XP_016865668.1 2 - 3 244 126
GenBank mRNA AF037448.1 - 2 - 3 672 126
AF155568.1 - 2 - 3 351 126
AK222776.1 - 2 - 3 297 126
AY034481.1 - 1 - 2 147 126
AY034482.1 - 1 - 2 147 126
AY034483.1 - 1 - 2 147 126
BC015575.2 - 2 - 3 229 126
BC024283.1 - 2 - 3 256 126
BC032643.1 - 2 - 3 244 126

View Full Product Details