Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNTG2
Entrez Gene ID
54221
Gene Name
syntrophin gamma 2
Gene Aliases
G2SYN, SYN5
Chromosome Location
Chr.2: 950868 - 1367991 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018968.3 NP_061841.2 10 - 11 982 73
XM_011510366.2 XP_011508668.1 8 - 9 1019 73
XM_017004361.1 XP_016859850.1 10 - 11 944 73
XM_017004362.1 XP_016859851.1 10 - 11 950 73
XM_017004363.1 XP_016859852.1 10 - 11 1131 73
XM_017004364.1 XP_016859853.1 7 - 8 4272 73
XM_017004365.1 XP_016859854.1 10 - 11 945 73
XM_017004366.1 XP_016859855.1 10 - 11 945 73
XM_017004367.1 XP_016859856.1 11 - 12 1020 73
XM_017004368.1 XP_016859857.1 10 - 11 945 73
XM_017004369.1 XP_016859858.1 10 - 11 945 73
XM_017004370.1 XP_016859859.1 10 - 11 945 73
XM_017004371.1 XP_016859860.1 10 - 11 946 73
XM_017004372.1 XP_016859861.1 10 - 11 946 73
XM_017004373.1 XP_016859862.1 5 - 6 1586 73
XM_017004374.1 XP_016859863.1 9 - 10 823 73
GenBank mRNA AJ003029.1 - 10 - 11 984 73
BC125251.1 - 4 - 5 530 73

View Full Product Details