Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNTG2
Entrez Gene ID
54221
Gene Name
syntrophin gamma 2
Gene Aliases
G2SYN, SYN5
Chromosome Location
Chr.2: 950868 - 1367991 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018968.3 NP_061841.2 2 - 3 338 98
XM_017004361.1 XP_016859850.1 2 - 3 300 98
XM_017004362.1 XP_016859851.1 2 - 3 306 98
XM_017004365.1 XP_016859854.1 2 - 3 301 98
XM_017004366.1 XP_016859855.1 2 - 3 301 98
XM_017004367.1 XP_016859856.1 2 - 3 338 98
XM_017004368.1 XP_016859857.1 2 - 3 301 98
XM_017004369.1 XP_016859858.1 2 - 3 301 98
XM_017004370.1 XP_016859859.1 2 - 3 301 98
XM_017004371.1 XP_016859860.1 2 - 3 302 98
XM_017004372.1 XP_016859861.1 2 - 3 302 98
XM_017004374.1 XP_016859863.1 2 - 3 307 98
GenBank mRNA AJ003029.1 - 2 - 3 340 98

View Full Product Details