The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018968.3 | NP_061841.2 | 4 - 5 | 454 | 95 |
XM_017004361.1 | XP_016859850.1 | 4 - 5 | 416 | 95 | |
XM_017004362.1 | XP_016859851.1 | 4 - 5 | 422 | 95 | |
XM_017004363.1 | XP_016859852.1 | 4 - 5 | 603 | 95 | |
XM_017004365.1 | XP_016859854.1 | 4 - 5 | 417 | 95 | |
XM_017004366.1 | XP_016859855.1 | 4 - 5 | 417 | 95 | |
XM_017004368.1 | XP_016859857.1 | 4 - 5 | 417 | 95 | |
XM_017004369.1 | XP_016859858.1 | 4 - 5 | 417 | 95 | |
XM_017004370.1 | XP_016859859.1 | 4 - 5 | 417 | 95 | |
XM_017004371.1 | XP_016859860.1 | 4 - 5 | 418 | 95 | |
XM_017004372.1 | XP_016859861.1 | 4 - 5 | 418 | 95 | |
XM_017004374.1 | XP_016859863.1 | 4 - 5 | 423 | 95 | |
GenBank mRNA | AJ003029.1 | - | 4 - 5 | 456 | 95 |