The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005592.3 | NP_005583.1 | 10 - 11 | 1492 | 99 |
XM_005251994.3 | XP_005252051.1 | 11 - 12 | 2104 | 99 | |
XM_011518708.2 | XP_011517010.1 | 2 - 3 | 237 | 99 | |
XM_017014734.1 | XP_016870223.1 | 10 - 11 | 1840 | 99 | |
GenBank mRNA | AF006464.1 | - | 10 - 11 | 1404 | 99 |