The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014675.4 | NP_055490.4 | 32 - 33 | 5367 | 56 |
XM_006711058.3 | XP_006711121.1 | 31 - 32 | 5158 | 56 | |
XM_011542468.1 | XP_011540770.1 | 32 - 33 | 5344 | 56 | |
XM_011542470.2 | XP_011540772.1 | 32 - 33 | 5344 | 56 | |
XM_017002913.1 | XP_016858402.1 | 32 - 33 | 5344 | 56 | |
XM_017002915.1 | XP_016858404.1 | 29 - 30 | 4852 | 56 | |
GenBank mRNA | AB007914.2 | - | 30 - 31 | 5006 | 56 |
AF527734.1 | - | 3 - 4 | 587 | 56 | |
BC126911.1 | - | 23 - 24 | 4507 | 56 | |
BC126912.1 | - | 23 - 24 | 4507 | 56 | |
DQ139275.1 | - | 19 - 20 | 4724 | 56 |