The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014675.4 | NP_055490.4 | 34 - 35 | 5724 | 74 |
XM_006711058.3 | XP_006711121.1 | 33 - 34 | 5515 | 74 | |
XM_011542468.1 | XP_011540770.1 | 34 - 35 | 5701 | 74 | |
XM_011542470.2 | XP_011540772.1 | 34 - 35 | 5701 | 74 | |
XM_017002913.1 | XP_016858402.1 | 34 - 35 | 5701 | 74 | |
XM_017002914.1 | XP_016858403.1 | 32 - 33 | 5326 | 74 | |
XM_017002915.1 | XP_016858404.1 | 31 - 32 | 5209 | 74 | |
GenBank mRNA | AB007914.2 | - | 32 - 33 | 5363 | 74 |
AF527734.1 | - | 5 - 6 | 944 | 74 | |
BC126911.1 | - | 25 - 26 | 4864 | 74 | |
BC126912.1 | - | 25 - 26 | 4864 | 74 | |
DQ139275.1 | - | 21 - 22 | 5081 | 74 |