Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MUSK
Entrez Gene ID
4593
Gene Name
muscle associated receptor tyrosine kinase
Gene Aliases
CMS9, FADS
Chromosome Location
Chr.9: 110668189 - 110806633 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166280.1 NP_001159752.1 3 - 4 483 143
NM_001166281.1 NP_001159753.1 3 - 4 483 143
NM_005592.3 NP_005583.1 3 - 4 483 143
XM_005251994.3 XP_005252051.1 3 - 4 1065 143
XM_005251995.3 XP_005252052.1 3 - 4 1064 143
XM_005251996.3 XP_005252053.1 3 - 4 1063 143
XM_017014734.1 XP_016870223.1 3 - 4 1065 143
GenBank mRNA AA448972.1 - 3 - 4 448 143
AF006464.1 - 3 - 4 395 143
BC109098.1 - 3 - 4 395 143
BC109099.1 - 3 - 4 395 143

View Full Product Details