Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RRNAD1
Entrez Gene ID
51093
Gene Name
ribosomal RNA adenine dimethylase domain containing 1
Gene Aliases
C1orf66, CGI-41, METTL25B
Chromosome Location
Chr.1: 156728471 - 156736960 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142560.1 NP_001136032.1 6 - 7 1460 75
NM_015997.3 NP_057081.3 7 - 8 1945 75
XM_005245235.4 XP_005245292.1 6 - 7 1589 75
XM_005245236.4 XP_005245293.1 7 - 8 1289 75
XM_011509600.1 XP_011507902.1 7 - 8 1282 75
XM_017001404.1 XP_016856893.1 7 - 8 1865 75
XM_017001405.1 XP_016856894.1 7 - 8 1810 75
XM_017001406.1 XP_016856895.1 7 - 8 1816 75
GenBank mRNA AF151800.1 - 7 - 8 1535 75
AK000412.1 - 8 - 9 1761 75
BX094595.1 - 1 - 2 292 75

View Full Product Details