The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015997.3 | NP_057081.3 | 5 - 6 | 1276 | 124 |
XM_005245234.4 | XP_005245291.1 | 5 - 6 | 1061 | 124 | |
XM_005245235.4 | XP_005245292.1 | 4 - 5 | 920 | 124 | |
XM_011509600.1 | XP_011507902.1 | 5 - 6 | 613 | 124 | |
XM_017001404.1 | XP_016856893.1 | 5 - 6 | 1196 | 124 | |
XM_017001405.1 | XP_016856894.1 | 5 - 6 | 1141 | 124 | |
XM_017001406.1 | XP_016856895.1 | 5 - 6 | 1147 | 124 | |
GenBank mRNA | AF151800.1 | - | 5 - 6 | 866 | 124 |
AK000412.1 | - | 6 - 7 | 1092 | 124 | |
BC011382.1 | - | 5 - 6 | 1261 | 124 |