The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127207.1 | NP_001120679.1 | 16 - 17 | 131 | |
NM_014140.3 | NP_054859.2 | 16 - 17 | 131 | ||
XM_005246631.2 | XP_005246688.1 | 16 - 17 | 131 | ||
XM_005246632.1 | XP_005246689.1 | 16 - 17 | 131 | ||
XM_017004228.1 | XP_016859717.1 | 16 - 17 | 131 | ||
GenBank mRNA | AF082179.1 | - | 15 - 16 | 131 | |
AF432223.1 | - | 16 - 17 | 131 | ||
AI701066.1 | - | 2 - 3 | 424 | 131 | |
AK000117.1 | - | 12 - 13 | 131 | ||
AK023780.1 | - | 16 - 17 | 131 | ||
AL122076.1 | - | 14 - 15 | 131 | ||
BC016482.1 | - | 15 - 16 | 131 | ||
BC029994.1 | - | 13 - 14 | 131 | ||
BC043341.1 | - | 16 - 17 | 131 |