Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPIL3
Entrez Gene ID
53938
Gene Name
peptidylprolyl isomerase like 3
Gene Aliases
CYPJ
Chromosome Location
Chr.2: 200870907 - 200889333 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032472.3 NP_115861.1 2 - 3 250 101
NM_130906.2 NP_570981.1 2 - 3 250 101
XM_005246651.3 XP_005246708.1 2 - 3 169 101
XM_005246652.4 XP_005246709.1 2 - 3 169 101
XM_011511358.2 XP_011509660.1 2 - 3 355 101
XM_017004352.1 XP_016859841.1 2 - 3 360 101
XM_017004353.1 XP_016859842.1 2 - 3 99 101
XM_017004354.1 XP_016859843.1 2 - 3 533 101
XM_017004355.1 XP_016859844.1 2 - 3 569 101
XM_017004356.1 XP_016859845.1 2 - 3 533 101
XM_017004357.1 XP_016859846.1 2 - 3 346 101
XM_017004358.1 XP_016859847.1 2 - 3 169 101
XM_017004359.1 XP_016859848.1 2 - 3 353 101
GenBank mRNA AF146799.1 - 2 - 3 119 101
AF251049.1 - 2 - 3 250 101
AF271652.1 - 2 - 3 185 101
AF414127.1 - 2 - 3 280 101
AK297804.1 - 2 - 3 181 101
AY257468.1 - 2 - 3 400 101
BC007693.2 - 2 - 3 691 101
CB961908.1 - 2 - 3 305 101

View Full Product Details