The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318060.1 | NP_001304989.1 | 3 - 4 | 897 | 138 |
NM_001318061.1 | NP_001304990.1 | 3 - 4 | 805 | 138 | |
NM_001318062.1 | NP_001304991.1 | 3 - 4 | 897 | 138 | |
NM_006716.3 | NP_006707.1 | 3 - 4 | 897 | 138 | |
GenBank mRNA | AB028069.1 | - | 3 - 4 | 910 | 138 |
AB028070.1 | - | 3 - 4 | 910 | 138 | |
AF160249.1 | - | 3 - 4 | 609 | 138 | |
AF160876.1 | - | 3 - 4 | 592 | 138 | |
AK292569.1 | - | 3 - 4 | 639 | 138 | |
AK302010.1 | - | 3 - 4 | 526 | 138 | |
AK303175.1 | - | 3 - 4 | 501 | 138 | |
BC022811.1 | - | 3 - 4 | 494 | 138 | |
BC036045.1 | - | 3 - 4 | 703 | 138 | |
BC047693.1 | - | 3 - 4 | 578 | 138 |