The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001907.2 | NP_001898.1 | 5 - 6 | 522 | 116 |
GenBank mRNA | AK223172.1 | - | 5 - 6 | 509 | 116 |
BC039716.1 | - | 5 - 6 | 514 | 116 | |
BC063475.1 | - | 5 - 6 | 500 | 116 | |
X71877.1 | - | 5 - 6 | 509 | 116 |