The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001257281.1 | NP_001244210.1 | 13 - 14 | 1858 | 64 |
GenBank mRNA | AK094293.1 | - | 13 - 14 | 1859 | 64 |