The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002118.4 | NP_002109.2 | 2 - 3 | 599 | 75 |
GenBank mRNA | AK295872.1 | - | 2 - 3 | 555 | 75 |
AK301017.1 | - | 2 - 3 | 326 | 75 | |
AK315979.1 | - | 2 - 3 | 532 | 75 | |
AY645722.1 | - | 2 - 3 | 569 | 75 | |
BC027175.1 | - | 2 - 3 | 480 | 75 | |
CR407606.1 | - | 2 - 3 | 336 | 75 | |
U15085.1 | - | 2 - 3 | 569 | 75 | |
Z23139.1 | - | 2 - 3 | 504 | 75 |