Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AP4E1
Entrez Gene ID
23431
Gene Name
adaptor related protein complex 4 epsilon 1 subunit
Gene Aliases
CPSQ4, SPG51, STUT1
Chromosome Location
Chr.15: 50908569 - 51005900 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252127.1 NP_001239056.1 3 - 4 478 120
NM_007347.4 NP_031373.2 3 - 4 455 120
XM_005254264.3 XP_005254321.1 3 - 4 273 120
XM_006720447.3 XP_006720510.1 3 - 4 633 120
XM_017022042.1 XP_016877531.1 3 - 4 400 120
GenBank mRNA AB030653.1 - 3 - 4 378 120
AF155156.2 - 3 - 4 387 120
AK297290.1 - 3 - 4 455 120
BC126308.1 - 3 - 4 454 120
BC130466.1 - 3 - 4 454 120
CR749604.1 - 3 - 4 355 120

View Full Product Details